missing translation for 'onlineSavingsMsg'
Learn More

4-Sulfamidobenzoyl chloride DMF complex, 95%

Product Code. 11354995
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11354995 5 g 5g
11344995 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11354995 Supplier Thermo Scientific Alfa Aesar Supplier No. L18428.06
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1172493-99-0 | C10H13ClN2O4S | 292.73 g/mol

It was used in the discovery of 2-Phenyl-3-sulfonylphenyl-indole derivatives as a new class of selective COX-2 inhibitors and in the synthesis and biological evaluation of substituted 2-sulfonyl-phenyl-3-phenyl-indoles: a new series of selective COX-2 inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It was used in the discovery of 2-Phenyl-3-sulfonylphenyl-indole derivatives as a new class of selective COX-2 inhibitors and in the synthesis and biological evaluation of substituted 2-sulfonyl-phenyl-3-phenyl-indoles: a new series of selective COX-2 inhibitors.

Solubility
Decomposes in water.

Notes
Moisture sensitive. Store away from water/moisture, strong bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1172493-99-0
Molecular Formula C10H13ClN2O4S
Molecular Weight (g/mol) 292.73
MDL Number MFCD02094019
InChI Key CRGFLVSVHAHYCI-UHFFFAOYSA-N
Synonym 4-sulfamidobenzoyl chloride dmf complex, 4-sulfamoylbenzoyl chloride; dimethylformamide, 4-sulphamido benzoyl chloride dmf complex, 4-aminosulfonyl benzoyl chloride dmf complex, 4-chlorocarbonyl benzenesulphonamide dmf complex
PubChem CID 44119145
SMILES CN(C)C=O.NS(=O)(=O)C1=CC=C(C=C1)C(Cl)=O

Specifications

Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Decomposes in water.
IUPAC Name N,N-dimethylformamide;4-sulfamoylbenzoyl chloride
Formula Weight 292.74
Percent Purity 95%
Chemical Name or Material 4-Sulfamidobenzoyl chloride DMF complex

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.