missing translation for 'onlineSavingsMsg'
Learn More

4-Sulfonamidophenylhydrazine hydrochloride, 97%

Product Code. 11440471
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Unit Size:
10g
2.5g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440471 50 g 50g
11420471 2.5 g 2.5g
11430471 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440471 Supplier Thermo Scientific Alfa Aesar Supplier No. B20015.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17852-52-7 | C6H10ClN3O2S | 223.675 g/mol

It is a potentially useful intermediate in the production of Celebrex an anti inflammatory drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a potentially useful intermediate in the production of Celebrex an anti inflammatory drug.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, water/moisture and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17852-52-7
Molecular Formula C6H10ClN3O2S
Molecular Weight (g/mol) 223.675
MDL Number MFCD00052262
InChI Key IKEURONJLPUALY-UHFFFAOYSA-N
Synonym 4-hydrazinylbenzenesulfonamide hydrochloride, 4-hydrazinobenzene-1-sulfonamide hydrochloride, 4-hydrazinobenzenesulfonamide hydrochloride, 4-sulfonamidephenylhydrazine hcl, 4-aminosulfonylphenylhydrazine hydrochloride, 4-hydrazinylbenzenesulfonamide hydrochloride 1:x, 4-sulfamoylphenyl hydrazine hydrochloride, 4-sulfonamide-phenylhydrazine hcl, phenylhydrazine-4-sulfonamide hydrochloride
PubChem CID 2794567
IUPAC Name 4-hydrazinylbenzenesulfonamide;hydrochloride
SMILES C1=CC(=CC=C1NN)S(=O)(=O)N.Cl

Specifications

Melting Point 217°C to 219°C (decomposition)
Quantity 50 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 223.68
Percent Purity 97%
Chemical Name or Material 4-Sulfonamidophenylhydrazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.