missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butyl-2,6-dinitrophenol, 97%

Product Code. 15448889
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15448889 25 g 25g
15438889 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448889 Supplier Thermo Scientific Alfa Aesar Supplier No. L08670.14
 Restricted Item
Estimated Shipment: 22-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4097-49-8 | C10H12N2O5 | 240.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4097-49-8
Molecular Formula C10H12N2O5
Molecular Weight (g/mol) 240.22
MDL Number MFCD00051969
InChI Key NJBDTWSOYUZQPM-UHFFFAOYSA-N
Synonym 4-tert-butyl-2,6-dinitrophenol, 2,6-dinitro-4-tert-butylphenol, unii-m5s512rl03, phenol, 4-1,1-dimethylethyl-2,6-dinitro, 2,6-dinitro-p-tert-butyl phenol, phenol, 4-tert-butyl-2,6-dinitro, acmc-1am7s, 2,6-dinitro-4-t-butylphenol, 4-t-butyl-2,6-dinitrophenol
PubChem CID 20042
IUPAC Name 4-tert-butyl-2,6-dinitrophenol
SMILES CC(C)(C)C1=CC(=C(O)C(=C1)[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 93°C to 95°C
Quantity 25 g
UN Number UN2811
Beilstein 1986466
Formula Weight 240.22
Percent Purity 97%
Chemical Name or Material 4-tert-Butyl-2,6-dinitrophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.