missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyl)benzene-1,2-diamine, 97%, Thermo Scientific™

Product Code. 10714947
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10714947 5 g 5g
10099281 250 mg 250mg
10631493 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10714947 Supplier Thermo Scientific Maybridge Supplier No. RF01562DE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 68176-57-8
Molecular Formula C10H16N2
Molecular Weight (g/mol) 164.252
MDL Number MFCD00052695
InChI Key WLOSFXSXVXTKBU-UHFFFAOYSA-N
PubChem CID 432708
IUPAC Name 4-tert-butylbenzene-1,2-diamine
SMILES CC(C)(C)C1=CC(=C(C=C1)N)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula (CH3)3CC6H3(NH2)2
Quantity 5 g
Synonym 4-tert-butyl benzene-1,2-diamine, 4-tert-butyl-1,2-diaminobenzene, 1,2-diamino-4-tert-butylbenzene, 4-tert-butyl-o-phenylenediamine, 1,2-benzenediamine,4-1,1-dimethylethyl, 1,2-benzenediamine, 4-1,1-dimethylethyl, pubchem22919, 4-tert-butyl-diaminobenzene, 4-t-butylbenzene-1,2-diamine, 4-tert-butylbenzen-1,2-diamine
Formula Weight 164.25
Percent Purity 97%
Chemical Name or Material 4-(tert-Butyl)benzene-1, 2-diamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.