missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyl)benzene-1-carbothioamide, ≥95%, Thermo Scientific™

Product Code. 10384242
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10384242 1 g 1g
10702693 10 g 10g
10782207 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10384242 Supplier Thermo Scientific Maybridge Supplier No. RF00754DA
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 57774-77-3
Molecular Formula C11H15NS
Molecular Weight (g/mol) 193.308
MDL Number MFCD00052485
InChI Key HZODWYBXBKXJLB-UHFFFAOYSA-N
Synonym 4-tert-butyl benzothioamide, 4-tert-butylthiobenzamide, 4-tert-butylbenzene-1-carbothioamide, 4-tert-butyl-thiobenzamide, 4-tert-butyl benzene-1-carbothioamide, amino 4-tert-butyl phenyl methane-1-thione, acmc-1az3y, benzenecarbothioamide,4-1,1-dimethylethyl, benzenecarbothioamide, 4-1,1-dimethylethyl-9ci
PubChem CID 2734817
IUPAC Name 4-tert-butylbenzenecarbothioamide
SMILES CC(C)(C)C1=CC=C(C=C1)C(=S)N

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Linear Formula (CH3)3CC6H4CSNH2
Quantity 1 g
Formula Weight 193.31
Percent Purity 95%
Chemical Name or Material 4-(tert-Butyl)benzene-1-carbothioamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.