missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyl)-o-phenylenediamine, 97%

Product Code. 15404897
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15404897 1 g 1g
15424897 250 mg 250mg
15414897 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15404897 Supplier Thermo Scientific Alfa Aesar Supplier No. H32205.03
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 68176-57-8 | C10H16N2 | 164.252 g/mol

4-(tert-Butyl)-o-phenylenediamine, is used as a as a co-catalyst for the polymerization of o-phenylenediamine and its derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(tert-Butyl)-o-phenylenediamine, is used as a as a co-catalyst for the polymerization of o-phenylenediamine and its derivatives.

Solubility
Soluble in most common organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68176-57-8
Molecular Formula C10H16N2
Molecular Weight (g/mol) 164.252
MDL Number MFCD00052695
InChI Key WLOSFXSXVXTKBU-UHFFFAOYSA-N
PubChem CID 432708
IUPAC Name 4-tert-butylbenzene-1,2-diamine
SMILES CC(C)(C)C1=CC(=C(C=C1)N)N

Specifications

Melting Point 98°C to 100°C
Quantity 1 g
UN Number UN2811
Sensitivity Air Sensitive
Synonym 4-tert-butyl benzene-1,2-diamine, 4-tert-butyl-1,2-diaminobenzene, 1,2-diamino-4-tert-butylbenzene, 4-tert-butyl-o-phenylenediamine, 1,2-benzenediamine,4-1,1-dimethylethyl, 1,2-benzenediamine, 4-1,1-dimethylethyl, pubchem22919, 4-tert-butyl-diaminobenzene, 4-t-butylbenzene-1,2-diamine, 4-tert-butylbenzen-1,2-diamine
Solubility Information Soluble in most common organic solvents.
Formula Weight 164.25
Percent Purity 97%
Chemical Name or Material 4-(tert-Butyl)-o-phenylenediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.