missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyl)phenyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 10662653
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10662653 1 g 1g
10743663 10 g 10g
10285464 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10662653 Supplier Thermo Scientific Maybridge Supplier No. TL00746DA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 19241-24-8
Molecular Formula C11H13NS
Molecular Weight (g/mol) 191.292
MDL Number MFCD00022059
InChI Key OCGNNCBNRBTUCG-UHFFFAOYSA-N
PubChem CID 140503
IUPAC Name 1-tert-butyl-4-isothiocyanatobenzene
SMILES CC(C)(C)C1=CC=C(C=C1)N=C=S

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula (CH3)3CC6H4NCS
Quantity 1 g
Synonym 4-tert-butylphenyl isothiocyanate, 4-t-butylphenyl isothiocyanate, benzene,1-1,1-dimethylethyl-4-isothiocyanato, 4-tert-butyl phenyl isothiocyanate, benzene, 1-1,1-dimethylethyl-4-isothiocyanato, 4-tert-butylphenylisothiocyanate, zlchem 964, 4-t-butylphenylisothiocyanate, p-t-butylphenyl isothiocyanate, 4-tert-butylphenyl-isothiocyanate
Formula Weight 191.3
Percent Purity 97%
Chemical Name or Material 4-(tert-Butyl)phenyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.