missing translation for 'onlineSavingsMsg'
Learn More

[4-(tert-Butyl)phenyl]methanesulfonyl chloride, Tech., Thermo Scientific™

Product Code. 10300603
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10300603 250 mg 250mg
10664073 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10300603 Supplier Thermo Scientific Maybridge Supplier No. MO00924CB
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 519056-61-2
Molecular Formula C11H15ClO2S
Molecular Weight (g/mol) 246.749
MDL Number MFCD04115413
InChI Key NEJDUPGGBSPJLG-UHFFFAOYSA-N
Synonym 4-tert-butylphenyl methanesulfonyl chloride, 4-tert-butyl phenyl methanesulfonyl chloride, 4-tert-butyl phenyl methanesulfonylchloride, 4-tert-butylphenyl methanesulphonyl chloride, 4-1-butyl phenylmethanesulfonyl chloride, 4-tert-butyl phenyl methyl chlorosulfone
PubChem CID 2794645
IUPAC Name (4-tert-butylphenyl)methanesulfonyl chloride
SMILES CC(C)(C)C1=CC=C(C=C1)CS(=O)(=O)Cl

Specifications

Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 246.76
Percent Purity Tech.
Grade Technical
Chemical Name or Material [4-(tert-Butyl)phenyl]methanesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.