missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylaniline, 99%

Product Code. 10439900 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10439900 5 mL 5mL
10072590 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10439900 Supplier Thermo Scientific Acros Supplier No. 198250050
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 769-92-6 | C10H15N | 149.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 769-92-6
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.24
MDL Number MFCD00007899
InChI Key WRDWWAVNELMWAM-UHFFFAOYSA-N
Synonym 4-tert-butyl aniline, p-tert-butylaniline, benzenamine, 4-1,1-dimethylethyl, p-t-butylaniline, 4-t-butylaniline, unii-47h9440zow, 4-t-butylbenzeneamine, 4-tert-butylbenzenamine, 4-tert-butylbenzeneamine
PubChem CID 69861
IUPAC Name 4-tert-butylaniline
SMILES CC(C)(C)C1=CC=C(C=C1)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 15°C to 16°C
Color Brown to Orange-Red
Density 0.9300g/mL
Boiling Point 230°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CC6H4NH2
Refractive Index 1.5370 to 1.539
Quantity 5 mL
Beilstein 12, 1166
Specific Gravity 0.93
Formula Weight 149.24
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-tert-Butylaniline, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-tert-Butylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 4-tert-Butylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.