missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzaldehyde, 95%

Product Code. 10624945 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10624945 250 mL Glass bottle 250mL
10376734 50 mL Glass bottle 50mL
10274914 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10624945 Supplier Thermo Scientific Acros Supplier No. 426522500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 939-97-9 | C11H14O | 162.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 939-97-9
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00035742
InChI Key OTXINXDGSUFPNU-UHFFFAOYSA-N
Synonym 4-tert-butyl benzaldehyde, p-t-butylbenzaldehyde, p-tertbutyl benzaldehyde, benzaldehyde, 4-1,1-dimethylethyl, p-tert-butylbenzaldehyde, 4-t-butylbenzaldehyde, 4-tert-butyl-benzaldehyde, unii-qxf0qy8503, 4-t-butyl benzaldehyde, benzaldehyde, p-tert-butyl
PubChem CID 70324
IUPAC Name 4-tert-butylbenzaldehyde
SMILES CC(C)(C)C1=CC=C(C=O)C=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9700g/mL
Boiling Point 249°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CC6H4CHO
Refractive Index 1.527
Quantity 250 mL
Specific Gravity 0.97
Solubility Information Solubility in water: insoluble
Viscosity 0.004 Pas (20°C)
Formula Weight 162.23
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-tert-Butylbenzaldehyde
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-tert-Butylbenzaldehyde
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.