missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzyl chloride, 99%

Product Code. 11413824
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413824 25 g 25g
11403824 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11413824 Supplier Thermo Scientific Alfa Aesar Supplier No. A11856.14
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 19692-45-6 | C11H15Cl | 182.69 g/mol

4-tert-Butylbenzyl chloride has been used to control molecular weight and prevent microgel formation during polymerization reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylbenzyl chloride has been used to control molecular weight and prevent microgel formation during polymerization reaction.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent. Store away from strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19692-45-6
Molecular Formula C11H15Cl
Molecular Weight (g/mol) 182.69
MDL Number MFCD00000918
InChI Key WAXIFMGAKWIFDQ-UHFFFAOYSA-N
Synonym 4-tert-butylbenzyl chloride, 1-tert-butyl-4-chloromethyl benzene, p-tert-butylbenzyl chloride, 4-tert-butyl benzyl chloride, benzene, 1-chloromethyl-4-1,1-dimethylethyl, 4-tert-butyl benzylchloride, p-t-butylbenzyl chloride, 1-chloromethyl-4-tert-butylbenzene, p-tert-butyl-.alpha.-chlorotoluene
PubChem CID 88198
IUPAC Name 1-tert-butyl-4-(chloromethyl)benzene
SMILES CC(C)(C)C1=CC=C(CCl)C=C1

Specifications

Density 0.945
Boiling Point 101°C to 104°C (7 mmHg)
Flash Point 94° (202°F)
Refractive Index 1.522
Quantity 25 g
UN Number UN1760
Beilstein 471673
Solubility Information Slightly soluble in water.
Formula Weight 182.7
Percent Purity 99%
Chemical Name or Material 4-tert-Butylbenzyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.