missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzylamine, 98%

Product Code. 11385659
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11385659

Brand: Thermo Scientific Alfa Aesar L20394.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39895-55-1 | C11H17N | 163.264 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Not miscible or difficult to mix with water.

Notes
Air Sensitive, store away form air. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39895-55-1
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD00040754
InChI Key MPWSRGAWRAYBJK-UHFFFAOYSA-N
Synonym 4-tert-butylbenzylamine, 4-tert-butylphenyl methanamine, 4-t-butylbenzylamine, 1-4-tert-butylphenyl methanamine, 4-tert-butyl-benzylamine, 4-tert-butyl phenyl methanamine, 4-tert-butyl phenyl methylamine, 4t-butylbenzylamine, p-t-butylbenzylamine, 4-tertbutylbenzylamine
PubChem CID 2735655
IUPAC Name (4-tert-butylphenyl)methanamine
SMILES CC(C)(C)C1=CC=C(C=C1)CN

Specifications

Density 0.927
Boiling Point 124°C (16 mmHg)
Flash Point 107°C (224°F)
Refractive Index 1.521
Quantity 5 g
UN Number UN2735
Beilstein 2961797
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 163.26
Percent Purity 98%
Chemical Name or Material 4-tert-Butylbenzylamine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.