missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butyldimethylsiloxy-1-butanol, 97%

Product Code. 15421498
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15421498 5 g 5g
15431498 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15421498 Supplier Thermo Scientific Alfa Aesar Supplier No. H55285.06
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 87184-99-4 | C10H24O2Si | 204.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly Soluble in water.(2.0 g/L) (25°C). Soluble in Dichloromethane, Ethyl Acetate, Methanol

Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87184-99-4
Molecular Formula C10H24O2Si
Molecular Weight (g/mol) 204.39
MDL Number MFCD04974070
InChI Key IJEMXJANZPVITP-UHFFFAOYSA-N
Synonym 4-tert-butyldimethylsilyl oxy butan-1-ol, 4-dimethyl-tert-butylsilyl oxybutan-1-ol, 4-dimethyl-tert-butylsilyloxy butan-1-ol, 4-tert-butyldimethylsilyl oxy-1-butanol, 1-butanol, 4-1,1-dimethylethyl dimethylsilyl oxy, acmc-20amup, 4-t-butyldimethylsiloxy-1-butanol, 4-t-butyldimethylsilyloxy-1-butanol, 4-tert-butyldimethylsiloxy-1-butanol
PubChem CID 186403
IUPAC Name 4-[tert-butyl(dimethyl)silyl]oxybutan-1-ol
SMILES CC(C)(C)[Si](C)(C)OCCCCO

Specifications

Boiling Point 63°C to 66°C (0.3mmHg)
Quantity 5 g
Linear Formula (CH3)3CSi(CH3)2O(CH2)4OH
Flash Point 105°C (221°F)
Solubility Information Slightly Soluble in water.(2.0g/L) (25°C). Soluble in Dichloromethane,Ethyl Acetate,Methanol
Formula Weight 204.38
Percent Purity 97%
Refractive Index 1.439
Density 0.888
Chemical Name or Material 4-tert-Butyldimethylsiloxy-1-butanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.