missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butyldimethylsiloxy-3-penten-2-one, 95%, mixture of isomers, Thermo Scientific™

Product Code. 11317489
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317489 5 g 5g
11327489 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317489 Supplier Thermo Scientific Alfa Aesar Supplier No. H27868.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69404-97-3
Molecular Formula C11H22O2Si
Molecular Weight (g/mol) 214.38
MDL Number MFCD00010655
InChI Key ZRVOCNOYAJIAAP-CSKARUKUSA-N
Synonym 2-t-butyldimethylsiloxy pent-2-en-4-one, 3e-4-tert-butyldimethylsilyl oxy pent-3-en-2-one, 4-tert-butyldimethylsiloxy-3-penten-2-one, 2-tert-butyldimethylsiloxy pent-2-en-4-one, 4-tert-butyldimethylsilyl oxy pent-3-en-2-one, acetylacetone enol tert-butyldimethylsilyl ether, 2-tert-butyldimethylsilyl oxy-2-penten-4-one, e-4-tert-butyldimethylsilyl oxy pent-3-en-2-one, e-4-tert-butyl dimethyl silyl oxypent-3-en-2-one, 4-tert-butyldimethylsiloxy-3-penten-2-one gc
PubChem CID 6034727
IUPAC Name (E)-4-[tert-butyl(dimethyl)silyl]oxypent-3-en-2-one
SMILES CC(=CC(=O)C)O[Si](C)(C)C(C)(C)C

Specifications

Boiling Point 50°C to 51°C (0.2mmHg)
Quantity 5 g
Flash Point 73°C (163°F)
Formula Weight 214.38
Percent Purity 95%
Sensitivity Moisture sensitive
Density 0.898
Chemical Name or Material 4-tert-Butyldimethylsiloxy-3-penten-2-one, mixture of isomers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.