missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylphenoxyacetic acid, 98%

Product Code. 11498866
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498866 5 g 5g
11408876 25 g 25g
11418876 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498866 Supplier Thermo Scientific Alfa Aesar Supplier No. A17017.06
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1798-04-5 | C12H16O3 | 208.257 g/mol

Applied as a reagent for the TAC protective group in nucleosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Applied as a reagent for the TAC protective group in nucleosides.

Solubility
Sparingly Soluble (0.56 g/L) (25°C), Calc.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1798-04-5
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.257
MDL Number MFCD00021758
InChI Key FBIGAJNVRFKBJL-UHFFFAOYSA-N
Synonym 4-tert-butylphenoxyacetic acid, 4-tert-butylphenoxy acetic acid, 4-tert-butyl-phenoxy-acetic acid, 2-4-tert-butylphenoxy acetic acid, p-tert-butylphenoxy acetic acid, p-t-butylphenoxyacetic acid, 2-4-tert-butyl phenoxy acetic acid, acetic acid, 4-1,1-dimethylethyl phenoxy, acetic acid, 2-4-1,1-dimethylethyl phenoxy
PubChem CID 15718
IUPAC Name 2-(4-tert-butylphenoxy)acetic acid
SMILES CC(C)(C)C1=CC=C(C=C1)OCC(=O)O

Specifications

Quantity 5 g
Solubility Information Sparingly Soluble (0.56g/L) (25°C),Calc.
Formula Weight 208.26
Percent Purity 98%
Chemical Name or Material 4-tert-Butylphenoxyacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.