missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylphenyl glycidyl ether, 95%

Product Code. 15432348
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15432348 25 g 25g
15422348 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15432348 Supplier Thermo Scientific Alfa Aesar Supplier No. H55945.14
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3101-60-8 | C13H18O2 | 206.285 g/mol

4-tert-Butylphenyl glycidyl ether is used in paint industry and serves as organic coating in Paint, boat Paint, building coating material. It is also used as an epoxy reactive diluent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylphenyl glycidyl ether is used in paint industry and serves as organic coating in Paint, boat Paint, building coating material. It is also used as an epoxy reactive diluent.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3101-60-8
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.285
MDL Number MFCD00005136
InChI Key HHRACYLRBOUBKM-UHFFFAOYSA-N
Synonym p-tert-butylphenyl glycidyl ether, 4-tert-butylphenyl glycidyl ether, 4-t-butylphenyl glycidyl ether, 2-4-tert-butylphenoxy methyl oxirane, tert-butylphenol glycidyl ether, tert-butylphenyl glycidyl ether, t-butyl phenyl glycidyl ether, ccris 2638, oxirane, 4-1,1-dimethylethyl phenoxy methyl, 2-4-tert-butyl phenoxy methyl oxirane
PubChem CID 18360
IUPAC Name 2-[(4-tert-butylphenoxy)methyl]oxirane
SMILES CC(C)(C)C1=CC=C(C=C1)OCC2CO2

Specifications

Color Colorless
Density 1.038 g/mL
Boiling Point 165°C to 170°C (14 mmHg)
Flash Point 102°C (216°F)
Refractive Index 1.515
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 206.28
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-tert-Butylphenyl glycidyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.