missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylphenylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™

Product Code. 10616470 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10616470 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10616470 Supplier Thermo Scientific Acros Supplier No. 432861000
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 63488-10-8 | C10H13BrMg | 237.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 63488-10-8, 60-29-7
CAS Min % 42.36, 53.19
CAS Max % 46.82, 57.64
Color Brown to Yellow
Density 1.0650g/mL
Flash Point −22°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C10H13BrMg
Linear Formula (CH3)3CC6H4MgBr
MDL Number MFCD00013244
Quantity 100 mL
Specific Gravity 1.065
Synonym 4-tert-butylphenylmagnesium bromide, 4-t-butylphenylmagnesium bromide, bromo 4-tert-butylphenyl magnesium, magnesium, bromo 4-1,1-dimethylethyl phenyl, 4-tert-butylphenylmagnesium bromide solution, yecfwfoeqlavnh-uhfffaoysa-m, 4-tert-butylphenyhnagnesium bromide, 4-tert.-butylphenylmagnesium bromide, 4-tert-butylphenyl magnesium bromide, 4-tert butyl phenyl magnesium bromide, grignard reagent
InChI Key YECFWFOEQLAVNH-UHFFFAOYSA-M
SMILES CC(C)(C)C1=CC=C([Mg]Br)C=C1
IUPAC Name magnesium;tert-butylbenzene;bromide
Molecular Weight (g/mol) 237.42
PubChem CID 4608371
Formula Weight 237.42
Physical Form Solution
Chemical Name or Material 4-tert-Butylphenylmagnesium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-tert-Butylphenylmagnesium bromide, 2M solution in diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P403+P235-Store in a well-ventilated place. Keep cool.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.