missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylthiobenzamide, 97%, Thermo Scientific™

Product Code. 15433487
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15433487 250 mg 250mg
11425920 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15433487 Supplier Thermo Scientific Alfa Aesar Supplier No. H26519.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57774-77-3
Molecular Formula C11H15NS
Molecular Weight (g/mol) 193.308
MDL Number MFCD00052485
InChI Key HZODWYBXBKXJLB-UHFFFAOYSA-N
Synonym 4-tert-butyl benzothioamide, 4-tert-butylthiobenzamide, 4-tert-butylbenzene-1-carbothioamide, 4-tert-butyl-thiobenzamide, 4-tert-butyl benzene-1-carbothioamide, amino 4-tert-butyl phenyl methane-1-thione, acmc-1az3y, benzenecarbothioamide,4-1,1-dimethylethyl, benzenecarbothioamide, 4-1,1-dimethylethyl-9ci
PubChem CID 2734817
IUPAC Name 4-tert-butylbenzenecarbothioamide
SMILES CC(C)(C)C1=CC=C(C=C1)C(=S)N

Specifications

Melting Point 145°C to 149°C
Quantity 250 mg
Formula Weight 193.31
Percent Purity 97%
Chemical Name or Material 4-tert-Butylthiobenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.