missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butyltoluene, 95%

Product Code. 15423177
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15423177 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15423177 Supplier Thermo Scientific Alfa Aesar Supplier No. A18214.AP
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 98-51-1 | C11H16 | 148.249 g/mol

4-tert-Butyltoluene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butyltoluene is used as pharmaceutical intermediate.

Solubility
Soluble in water (6 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 98-51-1
Melting Point -63°C
Density 0.853
Boiling Point 189°C to 193°C
Flash Point 62°C (144°F)
Odor Characteristic
Molecular Formula C11H16
Refractive Index 1.492
MDL Number MFCD00008837
Quantity 500 mL
UN Number UN2667
Beilstein 2038670
Synonym 4-tert-butyltoluene, p-tert-butyltoluene, 1-tert-butyl-4-methylbenzene, p-t-butyltoluene, benzene, 1-1,1-dimethylethyl-4-methyl, 4-t-butyltoluene, p-methyl-tert-butylbenzene, toluene, p-tert-butyl, 1-methyl-4-tert-butylbenzene, 4-methyl-tert-butylbenzene
Solubility Information Soluble in water (6g/L at 20°C).
InChI Key QCWXDVFBZVHKLV-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)C(C)(C)C
IUPAC Name 1-tert-butyl-4-methylbenzene
Molecular Weight (g/mol) 148.249
PubChem CID 7390
Formula Weight 148.25
Percent Purity 95%
Chemical Name or Material 4-tert-Butyltoluene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.