missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Melting Point | 281.0°C to 283.0°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 94% min. (HPLC) |
| Packaging | Glass Bottle |
| Quantity | 1 g |
| Formula Weight | 204.25 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-(Thien-3-yl)benzoic acid |
Safety and Handling
Product Identifier
- 4-(Thien-3-yl)benzoic acid, 97%
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?