missing translation for 'onlineSavingsMsg'
Learn More

4-Tolylsulfonamide, TRC

Product Code. 30132574
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132574 10 g 10g
17944595 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132574 Supplier Toronto Research Chemicals Supplier No. TRCT53663010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-55-3
Molecular Formula C7 H9 N O2 S
Molecular Weight (g/mol) 171.22
Synonym (4-Methylphenyl)sulfonamide, 4-Methylbenzenesulphonamide, Toluenesulfonamide, Tolbutamide Imp. A (EP), Gliclazide Imp. A (EP), Benzenesulfonamide, 4-methyl- (9CI, ACI), 4-Methylbenzenesulfonamide (ACI), p-Toluenesulfonamide (8CI), 4-Methylbenzene-1-sulfonamide, 4-Methylbenzensulfonamide, 4-Methylphenylsulfonamide, 4-Tolylsulfonamide, GWA 101, NSC 9908, p-Methylbenzenesulfonamide, p-Tolylsulfonamide, p-Tosylamide, Plasticizer 15, Toluene-4-sulfonamide, Tolylsulfonamide, Topcizer 1S, Tosylamide, Uniplex 173
IUPAC Name 4-methylbenzenesulfonamide
SMILES Cc1ccc(cc1)S(=O)(=O)N

Specifications

Quantity 10 g
InChI Formula InChI=1S/C7H9NO2S/c1-6-2-4-7(5-3-6)11(8,9)10/h2-5H,1H3,(H2,8,9,10)
Formula Weight 171.0354
Percent Purity >95%
Chemical Name or Material p-Toluenesulfonamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.