missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)aniline, 99%

Product Code. 10582651 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
25 g
Packaging:
Glass bottle
Unit Size:
2.5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10582651 25 g Glass bottle 25g
10165650 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10582651 Supplier Thermo Scientific Acros Supplier No. 297640250
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 461-82-5 | C7H6F3NO | 177.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 461-82-5
Molecular Formula C7H6F3NO
Molecular Weight (g/mol) 177.12
MDL Number MFCD00041314
InChI Key XUJFOSLZQITUOI-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy aniline, 4-trifluoromethoxy-phenylamine, 4-trifluoromethoxy benzenamine, p-trifluoromethoxyaniline, 4-trifluoroethoxy aniline, benzenamine, 4-trifluoromethoxy, unii-p40l42cvf6, alpha,alpha,alpha-trifluoro-p-anisidine, p-aminotrifluoromethoxybenzene, 4-amino-alpha,alpha,alpha-trifluoroanisole
PubChem CID 600848
IUPAC Name 4-(trifluoromethoxy)aniline
SMILES C1=CC(=CC=C1N)OC(F)(F)F

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.3100g/mL
Boiling Point 83.0°C to 85.0°C (15.0 mmHg)
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3OC6H4NH2
Refractive Index 1.4620 to 1.4640
Quantity 25 g
Specific Gravity 1.31
Formula Weight 177.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethoxy)aniline, 99+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-(Trifluoromethoxy)aniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.