missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzyl bromide, 97%

Product Code. 11493897
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11493897

Brand: Thermo Scientific Alfa Aesar A18335.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50824-05-0 | C8H6BrF3O | 255.034 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50824-05-0
Molecular Formula C8H6BrF3O
Molecular Weight (g/mol) 255.034
MDL Number MFCD00061238
InChI Key JDNPUJCKXLOHOW-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy benzyl bromide, 1-bromomethyl-4-trifluoromethoxy benzene, 4-trifluoromethoxybenzyl bromide, 4-trifluoromethoxy benzylbromide, p-trifluoromethoxy benzyl bromide, alpha-bromo-4-trifluoromethoxy toluene, benzene, 1-bromomethyl-4-trifluoromethoxy, 4-trifluoromethoxy-benzyl bromide, 4-bromomethyl phenoxy trifluoromethane, 1-bromomethyl-4-trifluoromethoxy-benzene
PubChem CID 142785
IUPAC Name 1-(bromomethyl)-4-(trifluoromethoxy)benzene
SMILES C1=CC(=CC=C1CBr)OC(F)(F)F

Specifications

Melting Point 22°C to 24°C
Density 1.594
Boiling Point 63°C (3 mmHg)
Flash Point 94°C (201°F)
Refractive Index 1.48
Quantity 25 g
UN Number UN3261
Beilstein 2521451
Formula Weight 255.04
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethoxy)benzyl bromide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.