missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzylamine, 97%, Thermo Scientific™

Product Code. 11459166
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459166 25 g 25g
11359449 1 g 1g
11449166 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459166 Supplier Thermo Scientific Alfa Aesar Supplier No. A18029.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93919-56-3
Molecular Formula C8H8F3NO
Molecular Weight (g/mol) 191.153
MDL Number MFCD00061237
InChI Key DBGROTRFYBSUTR-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy benzylamine, 4-trifluoromethoxy phenyl methanamine, 1-4-trifluoromethoxy phenyl methanamine, 4-trifluoromethoxybenzylamine, benzenemethanamine, 4-trifluoromethoxy, p-trifluoromethoxybenzylamine, p-trifluoromethoxy benzylamine, 4-trifluoromethoxy benzyl amine, 4-trifluoromethoxy phenyl methylamine
PubChem CID 571846
IUPAC Name [4-(trifluoromethoxy)phenyl]methanamine
SMILES C1=CC(=CC=C1CN)OC(F)(F)F

Specifications

Density 1.252
Boiling Point 57°C to 60°C (10 mmHg)
Flash Point 77°C (170°F)
Refractive Index 1.452
Quantity 25 g
UN Number UN2735
Beilstein 8200624
Sensitivity Air Sensitive
Formula Weight 191.15
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethoxy)benzylamine
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.