missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)phenol, 98%

Product Code. 10572091 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10572091 1 g Glass bottle 1g
10771735 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10572091 Supplier Thermo Scientific Acros Supplier No. 306040010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 828-27-3 | C7H5F3O2 | 178.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 828-27-3
Molecular Formula C7H5F3O2
Molecular Weight (g/mol) 178.11
MDL Number MFCD00040988
InChI Key WDRJNKMAZMEYOF-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy phenol, p-trifluoromethoxy phenol, phenol, 4-trifluoromethoxy, 4-hydroxytrifluoromethoxybenzene, 4-trifluoromethoxy-phenol, 4-hydroxyphenyl trifluoromethyl ether, p-trifluoromethoxyphenol
PubChem CID 70015
IUPAC Name 4-(trifluoromethoxy)phenol
SMILES OC1=CC=C(OC(F)(F)F)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown
Density 1.3700g/mL
Boiling Point 80.0°C (15.0 mmHg)
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3OC6H4OH
Refractive Index 1.4460 to 1.4480
Quantity 1 g
Specific Gravity 1.37
Formula Weight 178.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethoxy)phenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.