missing translation for 'onlineSavingsMsg'
Learn More

4'-(Trifluoromethyl)acetanilide, 98+%

Product Code. 11405143
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405143 5 g 5g
11415143 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11405143 Supplier Thermo Scientific Alfa Aesar Supplier No. A12789.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 349-97-3 | C9H8F3NO | 203.164 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 349-97-3
Molecular Formula C9H8F3NO
Molecular Weight (g/mol) 203.164
MDL Number MFCD00013562
InChI Key DFDHFECLWHHELH-UHFFFAOYSA-N
Synonym 4-trifluoromethyl acetanilide, n-4-trifluoromethyl phenyl acetamide, 4'-trifluoromethyl acetanilide, 4-trifluoromethylacetanilide, p-acetaminobenzotrifluoride, p-acetamidobenzotrifluoride, p-trifluoromethylacetanilide, p-acetylaminobenzotrifluoride, p-trifluoromethyl acetanilide
PubChem CID 67685
IUPAC Name N-[4-(trifluoromethyl)phenyl]acetamide
SMILES CC(=O)NC1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 150°C to 154°C
Odor Odorless
Quantity 5 g
Beilstein 2106965
Formula Weight 203.16
Percent Purity ≥98%
Chemical Name or Material 4'-(Trifluoromethyl)acetanilide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.