missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)aniline, ≥97%, Thermo Scientific™

Product Code. 10075853
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10075853 10 g 10g
10516334 1 g 1g
10381084 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10075853 Supplier Thermo Scientific Maybridge Supplier No. SB00719EA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 455-14-1
Molecular Formula C7H6F3N
Molecular Weight (g/mol) 161.127
MDL Number MFCD00064396
InChI Key ODGIMMLDVSWADK-UHFFFAOYSA-N
Synonym 4-trifluoromethyl aniline, 4-aminobenzotrifluoride, p-aminobenzotrifluoride, p-trifluoromethylaniline, benzenamine, 4-trifluoromethyl, 4-trifluoromethyl benzenamine, p-trifluoromethyl aniline, aniline, p-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-toluidine, a,a,a-trifluoro-p-toluidine
PubChem CID 9964
ChEBI CHEBI:40750
IUPAC Name 4-(trifluoromethyl)aniline
SMILES C1=CC(=CC=C1C(F)(F)F)N

Specifications

CAS Min % 97.0
Density 1.2960g/mL
Assay Percent Range 97+%
Packaging Amber Glass Bottle
Linear Formula H2NC6H4CF3
Quantity 10 g
Formula Weight 161.13
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)aniline
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Aminobenzotrifluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 4-Aminobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.