missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzene-1-carbothioamide, 97%, Thermo Scientific™

Product Code. 10389292
Change view
Click to view available options
Quantity:
25 g
Packaging:
Amber Glass Bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10389292 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10389292 Supplier Thermo Scientific Maybridge Supplier No. RF03219EB
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 72505-21-6
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H6F3NS
Linear Formula CF3C6H4CSNH2
MDL Number MFCD00051806
Quantity 25 g
Synonym 4-trifluoromethyl thiobenzamide, 4-trifluoromethyl benzenecarbothioamide, 4-trifluoromethylthiobenzamide, 4-trifluoromethyl benzene-1-carbothioamide, 4-trifluoromethyl benzothioamide, 4-trifluoromethyl-thiobenzamide, benzenecarbothioamide, 4-trifluoromethyl, 4-trifluoromethyl thiobenzamine
InChI Key IPRFNMJROWWFBH-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C(=S)N)C(F)(F)F
IUPAC Name 4-(trifluoromethyl)benzenecarbothioamide
Molecular Weight (g/mol) 205.198
PubChem CID 2734823
Formula Weight 205.2
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)benzene-1-carbothioamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.