missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzene-1-carboximidamide hydrochloride dihydrate, 97%, Thermo Scientific™

Product Code. 10596795
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10596795 5 g 5g
10436872 1 g 1g
10203432 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10596795 Supplier Thermo Scientific Maybridge Supplier No. SPB01314DE
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Identifiants chimiques

CAS 175278-62-3
Molecular Formula C8H12ClF3N2O2
Molecular Weight (g/mol) 260.64
MDL Number MFCD00122370
InChI Key ZFGZRQQZWBYRRB-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzamidine hydrochloride dihydrate, 4-trifluoromethyl benzimidamide hydrochloride dihydrate, 4-trifluoromethyl benzene-1-carboximidamide hydrochloride dihydrate, 4-trifluoromethyl benzenecarboximidamide dihydrate hydrochloride, benzenecarboximidamide,4-trifluoromethyl-, hydrochloride, hydrate 1:1:2, 4-trifluoromethyl benzamidine hydrochloridedihydrate, 4-trifluoromethylbenzamidine hydrochloride dihydrate, 4-trifluoromethyl benzamidine hydrochloride,dihydrate, 4-trifluoromethyl benzene-1-carboximidamide dihydrate hydrochloride, 4-trifluoromethyl benzene-1-carboximidamide-hydrogen chloride-water 1/1/2
PubChem CID 44630771
SMILES O.O.Cl.NC(=N)C1=CC=C(C=C1)C(F)(F)F

Spécification

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula HCl·H2O
Quantity 5 g
IUPAC Name 4-(trifluoromethyl)benzenecarboximidamide;dihydrate;hydrochloride
Formula Weight 242.63
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)benzene-1-carboximidamide hydrochloride hydrate
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.