missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzenepropanamine, TRC

Product Code. 30454035
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30454035 100 mg 100mg
30454246 250 mg 250mg
30452685 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30454035 Supplier Toronto Research Chemicals Supplier No. TRCT896343100MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101488-60-2
Molecular Formula C10H12NF3
Molecular Weight (g/mol) 203.21
Synonym 3-(4-Trifluoromethylphenyl)-1-propylamine, 3-[4-(Trifluoromethyl)phenyl]propylamine
IUPAC Name 3-[4-(trifluoromethyl)phenyl]propan-1-amine
SMILES NCCCc1ccc(cc1)C(F)(F)F

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H12F3N/c11-10(12,13)9-5-3-8(4-6-9)2-1-7-14/h3-6H,1-2,7,14H2
Formula Weight 203.0922
Chemical Name or Material 4-(Trifluoromethyl)benzenepropanamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.