missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzoic acid, 97%, Maybridge

Product Code. 10215904
Change view
Click to view available options
25g; 25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10215904 25g; 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10215904 Supplier Thermo Scientific Maybridge Supplier No. SB00672EB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 455-24-3
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H5F3O2
Linear Formula CF3C6H4CO2H
Quantity 25 g
Synonym 4-trifluoromethyl benzoic acid, p-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-p-toluic acid, benzoic acid, 4-trifluoromethyl, p-carboxybenzotrifluoride, 4-trifluoromethyl-benzoic acid, a,a,a-trifluoro-p-toluic acid, ptf-boa, 4-carboxybenzotrifluoride
InChI Key SWKPKONEIZGROQ-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C(=O)O)C(F)(F)F
IUPAC Name 4-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol) 190.121
PubChem CID 9966
ChEBI CHEBI:60696
Formula Weight 190.12
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)benzoic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.