missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzoic acid, 98+%

Product Code. 11481828
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11481828 5 g 5g
11491828 25 g 25g
11401838 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11481828 Supplier Thermo Scientific Alfa Aesar Supplier No. A14494.06
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 455-24-3 | C8H5F3O2 | 190.121 g/mol

4-(Trifluoromethyl)benzoic acid was used in the synthesis of salicylanilide 4-(trifluoromethyl)benzoates via N,N?-dicyclohexylcarbodiimide coupling in dry N,N-dimethylformamide. It was used as internal standard during the ultra trace analysis of fluorinated aromatic carboxylic acids by GC/MS method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)benzoic acid was used in the synthesis of salicylanilide 4-(trifluoromethyl)benzoates via N,N′-dicyclohexylcarbodiimide coupling in dry N,N-dimethylformamide. It was used as internal standard during the ultra trace analysis of fluorinated aromatic carboxylic acids by GC/MS method.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 455-24-3
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.121
MDL Number MFCD00002562
InChI Key SWKPKONEIZGROQ-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzoic acid, p-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-p-toluic acid, benzoic acid, 4-trifluoromethyl, p-carboxybenzotrifluoride, 4-trifluoromethyl-benzoic acid, a,a,a-trifluoro-p-toluic acid, ptf-boa, 4-carboxybenzotrifluoride
PubChem CID 9966
ChEBI CHEBI:60696
IUPAC Name 4-(trifluoromethyl)benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)C(F)(F)F

Specifications

Melting Point 220°C to 222°C
Boiling Point 247°C (753 mmHg)
Quantity 5 g
Beilstein 2049241
Solubility Information Soluble in water.
Formula Weight 190.12
Percent Purity ≥98%
Chemical Name or Material 4-(Trifluoromethyl)benzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.