Learn More
4-(Trifluoromethyl)benzoic acid, 98+%
Description
4-(Trifluoromethyl)benzoic acid was used in the synthesis of salicylanilide 4-(trifluoromethyl)benzoates via N,N?-dicyclohexylcarbodiimide coupling in dry N,N-dimethylformamide. It was used as internal standard during the ultra trace analysis of fluorinated aromatic carboxylic acids by GC/MS method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-(Trifluoromethyl)benzoic acid was used in the synthesis of salicylanilide 4-(trifluoromethyl)benzoates via N,N′-dicyclohexylcarbodiimide coupling in dry N,N-dimethylformamide. It was used as internal standard during the ultra trace analysis of fluorinated aromatic carboxylic acids by GC/MS method.
Solubility
Soluble in water.
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 220°C to 222°C |
| Boiling Point | 247°C (753 mmHg) |
| Quantity | 25 g |
| Beilstein | 2049241 |
| Solubility Information | Soluble in water. |
| Formula Weight | 190.12 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 4-(Trifluoromethyl)benzoic acid |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.