missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzyl alcohol, 99%

Product Code. 10256650 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10256650 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10256650 Supplier Thermo Scientific Acros Supplier No. 213570050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 349-95-1 | C8H7F3O | 176.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 349-95-1
CAS Min % 98.5
CAS Max % 100.0
Melting Point 18.0°C to 20.0°C
Color Colorless to Brown
Density 1.2800g/mL
Boiling Point 78.0°C to 80.0°C (4.0 mmHg)
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H7F3O
Linear Formula CF3C6H4CH2OH
Refractive Index 1.4590 to 1.4610
MDL Number MFCD00004661
Quantity 5 g
Specific Gravity 1.28
Synonym 4-trifluoromethyl benzyl alcohol, 4-trifluoromethyl phenyl methanol, p-trifluoromethylbenzyl alcohol, 4-trifluoromethyl benzene methanol, 4-trifluoromethyl benzylic alcohol, benzenemethanol, 4-trifluoromethyl, 4-trifluoromethylbenzyl alcohol, ccris 5117, 4-trifluoromethyl benzylalcohol
InChI Key MOOUWXDQAUXZRG-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1CO)C(F)(F)F
IUPAC Name [4-(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol) 176.14
PubChem CID 67684
Formula Weight 176.14
Percent Purity 99%
Physical Form Liquid After Melting
Chemical Name or Material 4-(Trifluoromethyl)benzyl alcohol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Trifluoromethyl)benzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.