missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzyl bromide, 98%

Codice prodotto. 11422928
Cambia vista
Click to view available options
Quantity:
5 g
25 g
Dimensione della confezione:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Dimensione della confezione
11422928 25 g 25g
11412928 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11422928 Fornitore Thermo Scientific Alfa Aesar N. del fornitore A16895.14
 Restricted Item
In Stock
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 402-49-3 | C8H6BrF3 | 239.035 g/mol

4-(Trifluoromethyl)benzyl bromide can be used to produce p-trifluoromethylbenzyl phenyl sulfide. It is used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)benzyl bromide can be used to produce p-trifluoromethylbenzyl phenyl sulfide. It is used in medicine.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 402-49-3
Molecular Formula C8H6BrF3
Molecular Weight (g/mol) 239.035
MDL Number MFCD00000403
InChI Key IKSNDOVDVVPSMA-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzyl bromide, 1-bromomethyl-4-trifluoromethyl benzene, 4-trifluoromethylbenzyl bromide, 4-bromomethyl benzotrifluoride, p-trifluoromethylbenzyl bromide, 4-trifluoromethyl benzylbromide, benzene, 1-bromomethyl-4-trifluoromethyl, 4-trifluoromethylbenzylbromide, p-trifluoromethyl benzyl bromide, alpha'-bromo-alpha,alpha,alpha-trifluoro-p-xylene
PubChem CID 123062
IUPAC Name 1-(bromomethyl)-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1CBr)C(F)(F)F

Specifica

Melting Point 29°C to 33°C
Density 1.546
Boiling Point 65°C to 66°C (5 mmHg)
Flash Point 88°C (190°F)
Odor Pungent
Refractive Index 1.4915
Quantity 25 g
UN Number UN3261
Beilstein 638980
Solubility Information Insoluble in water.
Formula Weight 239.04
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)benzyl bromide
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.