missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzylamine, 97%

Product Code. 10746573 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10746573 10 g Glass bottle 10g
10200513 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10746573 Proveedor Thermo Scientific Acros N.º de proveedor 375830100
 Restricted Item
In Stock
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 3300-51-4 | C8H8F3N | 175.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 3300-51-4
Molecular Formula C8H8F3N
Molecular Weight (g/mol) 175.15
MDL Number MFCD00010220
InChI Key PRDBLLIPPDOICK-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzylamine, 4-trifluoromethylbenzylamine, 4-trifluoromethyl phenyl methanamine, 1-4-trifluoromethyl phenyl methanamine, 4-trifluoromethyl benzyl amine, p-trifluoromethylbenzyl amine, 4-trifluoromethyl-benzylamine, p-trifluoromethyl benzylamine, benzenemethanamine, 4-trifluoromethyl
PubChem CID 76804
IUPAC Name [4-(trifluoromethyl)phenyl]methanamine
SMILES NCC1=CC=C(C=C1)C(F)(F)F

Especificaciones

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2290g/mL
Boiling Point 79°C to 82°C (15.0 mmHg)
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CF3C6H4CH2NH2
Refractive Index 1.4630 to 1.465
Quantity 10 g
Specific Gravity 1.229
Formula Weight 175.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethyl)benzylamine
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • 4-(Trifluoromethyl)benzylamine
Palabra de advertencia
  • Danger
Clasificación
  • Skin corrosion/irritation Category 1 B
Indicaciones de peligro
  • H314-Causes severe skin burns and eye damage.
Consejos de prudencia
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.