missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)phenylglyoxal hydrate, 98%, dry wt. basis

Product Code. 11329146
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11329146 5 g 5g
11339146 250 mg 250mg
11319146 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11329146 Supplier Thermo Scientific Alfa Aesar Supplier No. L20275.06
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1736-56-7 | C9H5F3O2 | 202.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1736-56-7
Molecular Formula C9H5F3O2
Molecular Weight (g/mol) 202.132
MDL Number MFCD05664098
InChI Key BGOMXTCPIUNFKR-UHFFFAOYSA-N
Synonym 2-oxo-2-4-trifluoromethyl phenyl acetaldehyde, 2-oxo-2-4-trifluoromethyl phenyl ethanal, 2-oxidanylidene-2-4-trifluoromethyl phenyl ethanal, 4-trifluoromethylphenyl oxo acetaldehyde, oxo 4-trifluoromethyl phenyl acetaldehyde, 1-4-trifluoromethyl phenyl-1,2-ethanedione
PubChem CID 2783286
IUPAC Name 2-oxo-2-[4-(trifluoromethyl)phenyl]acetaldehyde
SMILES C1=CC(=CC=C1C(=O)C=O)C(F)(F)F

Specifications

Melting Point 83°C to 85°C
Quantity 5 g
Beilstein 2261131
Formula Weight 202.13 (Anhydrous)
Percent Purity 98%
Assay (dry wt.)
Chemical Name or Material 4-(Trifluoromethyl)phenylglyoxal hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.