missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)phenylhydrazine, 96%

Product Code. 10746085 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10746085 1 g Glass bottle 1g
10516504 5 g Glass bottle 5g
2 options
This item is not returnable. View return policy

Product Code. 10746085

Brand: Thermo Scientific Acros 397950010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 368-90-1 | C7H7F3N2 | 176.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 368-90-1
Molecular Formula C7H7F3N2
Molecular Weight (g/mol) 176.14
MDL Number MFCD00042508
InChI Key DBNLGTYGKCMLLR-UHFFFAOYSA-N
Synonym 4-trifluoromethyl phenylhydrazine, 4-trifluoromethyl phenyl hydrazine, 4-trifluoromethylphenylhydrazine, p-trifluoromethylphenylhydrazine, hydrazine, 4-trifluoromethyl phenyl, p-trifluoromethyl phenylhydrazine, 4-trifluoromethyl-phenylhydrazine, 4-trifluoromethyl-phenyl-hydrazine
PubChem CID 136200
IUPAC Name [4-(trifluoromethyl)phenyl]hydrazine
SMILES NNC1=CC=C(C=C1)C(F)(F)F

Especificaciones

CAS Min % 95.0
CAS Max % 100.0
Melting Point 59°C to 64°C
Color Yellow to Brown
Boiling Point 118°C to 122°C (17.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula CF3C6H4NHNH2
Quantity 1 g
Formula Weight 176.14
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 4-(Trifluoromethyl)phenylhydrazine
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • 4-(Trifluoromethyl)phenylhydrazine
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.