missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)pyridine, 97%

Product Code. 11410028
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410028 1 g 1g
11420028 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410028 Supplier Thermo Scientific Alfa Aesar Supplier No. B22951.03
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 3796-24-5 | C6H4F3N | 147.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3796-24-5
Molecular Formula C6H4F3N
Molecular Weight (g/mol) 147.1
MDL Number MFCD00153289
InChI Key IIYVNMXPYWIJBL-UHFFFAOYSA-N
Synonym 4-trifluoromethyl pyridine, pyridine, 4-trifluoromethyl, 4-trifluoromethyl-pyridine, pubchem8028, p-trifluoromethylpyridine, acmc-1ctve
PubChem CID 138055
IUPAC Name 4-(trifluoromethyl)pyridine
SMILES C1=CN=CC=C1C(F)(F)F

Specifications

Density 1.27
Boiling Point 107°C to 110°C
Flash Point 20°C (68°F)
Refractive Index 1.4147
Quantity 1 g
UN Number UN1993
Beilstein 1364589
Formula Weight 147.1
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)pyridine
compliance-icons compliance-icons
Product Identifier
  • 4-(Trifluoromethyl)pyridine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4-(Trifluoromethyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.