missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)thiobenzamide, 98%, Thermo Scientific™

Product Code. 10587303 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
Product Code. Quantity Packaging unitSize
10587303 1 g Glass bottle 1g
1 options
This item is not returnable. View return policy

Product Code. 10587303

Brand: Thermo Scientific 374400010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 72505-21-6
Melting Point 136°C to 137°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C8H6F3NS
Linear Formula CF3C6H4CSNH2
Quantity 1 g
Synonym 4-trifluoromethyl thiobenzamide, 4-trifluoromethyl benzenecarbothioamide, 4-trifluoromethylthiobenzamide, 4-trifluoromethyl benzene-1-carbothioamide, 4-trifluoromethyl benzothioamide, 4-trifluoromethyl-thiobenzamide, benzenecarbothioamide, 4-trifluoromethyl, 4-trifluoromethyl thiobenzamine
InChI Key IPRFNMJROWWFBH-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C(=S)N)C(F)(F)F
IUPAC Name 4-(trifluoromethyl)benzenecarbothioamide
Molecular Weight (g/mol) 205.2
PubChem CID 2734823
Formula Weight 205.2
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)thiobenzamide
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Trifluoromethyl)thiobenzamide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.