missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)thiophenol, Technical Grade, Maybridge

Product Code. 10193764
Change view
Click to view available options
:
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10193764 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10193764 Supplier Thermo Scientific Maybridge Supplier No. TL00292DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 825-83-2
Packaging Amber Glass Bottle
Molecular Formula C7H5F3S
Quantity 1 g
Synonym 4-trifluoromethyl thiophenol, 4-trifluoromethyl benzenethiol, 4-trifluoromethyl benzene-1-thiol, 4-trifluoromethylthiophenol, 4-mercaptobenzotrifluoride, benzenethiol, 4-trifluoromethyl, 4-trifluoromethyl t, acmc-20amtl, pubchem6864
InChI Key WCMLRSZJUIKVCW-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C(F)(F)F)S
IUPAC Name 4-(trifluoromethyl)benzenethiol
Molecular Weight (g/mol) 178.172
PubChem CID 136653
Formula Weight 178.18
Percent Purity Tech.
Grade Technical
Chemical Name or Material 4-(Trifluoromethyl)benzene-1-thiol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.