missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethylsulfonyl)benzonitrile, 97+%

Product Code. 11439957
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439957 1 g 1g
11449957 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439957 Supplier Thermo Scientific Alfa Aesar Supplier No. B22907.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 312-21-0 | C8H4F3NO2S | 235.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 312-21-0
Molecular Formula C8H4F3NO2S
Molecular Weight (g/mol) 235.18
MDL Number MFCD01631631
InChI Key UHCGSMMNICMDIX-UHFFFAOYSA-N
Synonym 4-trifluoromethylsulfonyl benzonitrile, 4-trifluoromethyl sulfonyl benzonitrile, 4-trifluoromethanesulfonyl benzonitrile, 4-trifluoromethanesulfonylbenzonitrile, 4-trifluoromethyl sulfonyl benzenecarbonitrile, acmc-1clj3, 4-trifluoromethanesulphonylbenzonitrile, 4-trifluoromethylsulfonyl benzonitril
PubChem CID 2777824
IUPAC Name 4-(trifluoromethylsulfonyl)benzonitrile
SMILES C1=CC(=CC=C1C#N)S(=O)(=O)C(F)(F)F

Specifications

Melting Point 87°C to 92°C
Quantity 1 g
UN Number UN3439
Formula Weight 235.18
Percent Purity ≥97%
Chemical Name or Material 4-(Trifluoromethylsulfonyl)benzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.