missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethylthio)aniline, 98%

Product Code. 11352877
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352877 1 g 1g
11362877 5 g 5g
11372877 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11352877 Supplier Thermo Scientific Alfa Aesar Supplier No. L16067.03
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 372-16-7 | C7H6F3NS | 193.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372-16-7
Molecular Formula C7H6F3NS
Molecular Weight (g/mol) 193.19
MDL Number MFCD00040926
InChI Key OHHHTUXVBNGOGI-UHFFFAOYSA-N
Synonym 4-trifluoromethylthio aniline, 4-trifluoromethyl thio aniline, 4-trifluoromethyl sulfanyl aniline, 4-aminophenyl trifluoromethyl sulfide, 4-aminophenyl trifluoromethyl sulphide, 4-trifluoromethylsulfanyl aniline, benzenamine, 4-trifluoromethyl thio, 4-trifluoromethylthio phenylamine
PubChem CID 123054
SMILES NC1=CC=C(SC(F)(F)F)C=C1

Specifications

Density 1.344
Boiling Point 103°C (11 mmHg)
Flash Point 106°C (222°F)
Odor Unpleasant
Refractive Index 1.528
Quantity 1 g
UN Number UN2810
Beilstein 2208859
IUPAC Name 4-(trifluoromethylsulfanyl)aniline
Formula Weight 193.19
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethylthio)aniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.