missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethylthio)phenylacetic acid, 98%

Product Code. 15490119
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490119 1 g 1g
15410129 250 mg 250mg
15400129 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490119 Supplier Thermo Scientific Alfa Aesar Supplier No. H63432.03
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 243977-23-3 | C9H7F3O2S | 236.208 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 243977-23-3
Molecular Formula C9H7F3O2S
Molecular Weight (g/mol) 236.208
MDL Number MFCD00236351
InChI Key BFDKCZMYQOSTJG-UHFFFAOYSA-N
Synonym 4-trifluoromethylthio phenylacetic acid, 2-4-trifluoromethyl thio phenyl acetic acid, benzeneacetic acid, 4-trifluoromethyl thio, 4-trifluoromethylsulfanyl-phenyl-acetic acid, 2-4-trifluoromethylthio phenyl acetic acid, 4-trifluoromethyl sulfanyl phenyl acetic acid, pubchem7296, acmc-1chv0
PubChem CID 2777902
IUPAC Name 2-[4-(trifluoromethylsulfanyl)phenyl]acetic acid
SMILES C1=CC(=CC=C1CC(=O)O)SC(F)(F)F

Specifications

Quantity 1 g
Formula Weight 236.21
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-(Trifluoromethylthio)phenylacetic acid

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.