missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylbenzyl chloride, 90%, technical

Product Code. 10013443 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10013443 500 g Glass bottle 500g
10403053 25 g Glass bottle 25g
10770682 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10013443 Supplier Thermo Scientific Acros Supplier No. 422525000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1592-20-7 | C9H9Cl | 152.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1592-20-7
Molecular Formula C9H9Cl
Molecular Weight (g/mol) 152.62
MDL Number MFCD00051362
InChI Key ZRZHXNCATOYMJH-UHFFFAOYSA-N
Synonym 4-vinylbenzyl chloride, 1-chloromethyl-4-vinylbenzene, 4-chloromethylstyrene, 4-vinylbenzylchloride, benzene, 1-chloromethyl-4-ethenyl, 1-chloromethyl-4-ethenylbenzene, 4-chloromethyl styrene, p-vinylbenzyl chloride, unii-cjl4oj6bs0, cjl4oj6bs0
PubChem CID 74126
IUPAC Name 1-(chloromethyl)-4-ethenylbenzene
SMILES ClCC1=CC=C(C=C)C=C1

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Yellow
Density 1.0800g/mL
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula CH2CHC6H4CH2Cl
Refractive Index 1.5731 to 1.5751
Quantity 500 g
Specific Gravity 1.08
Stabilizer about 500ppm 4-tert-Butylcatechol, about 500ppm Nitrocresol + Nitrophenol
Formula Weight 152.62
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material 4-Vinylbenzyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.