missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Vinylphenol, 95%, 10% solution in propylene glycol, Thermo Scientific Chemicals
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifikationer
| CAS Min % | 89.0, 9.0 |
| CAS Max % | 91.0, 11.0 |
| Color | Colorless to Orange |
| Density | 1.0390g/mL |
| Boiling Point | 189°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 9 to 15% (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.4420 to 1.4520 (20°C, 589nm) |
| Quantity | 25 mL |
| Visa mer |
Säkerhet och hantering
Produktbeteckning
- 4-Vinylphenol, 10% solution in propylene glycol
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?