Learn More
RETSCH 400 dia. x 65mmH Stainless Steel Test Sieve, ISO Certified
Brand:  RETSCH 60.166.000125
Additional Details : Weight : 1.60000kg
 View more versions of this product
Product Code. 10620492
  • 296.00 € / Each
Estimated Shipment: 16-07-2024
to see stock.
Add to basket
Specifications
Stainless Steel
Stainless Steel
1Each
Test Sieve
Stainless Steel
65mm
For separation, fractioning, particle size determination
400 mm
125μm
ISO 3310/1
Shaker AS 450