missing translation for 'onlineSavingsMsg'
Learn More

42-O-tert-Butyldimethylsilyloxyethyl Rapamycin, TRC

Product Code. 30007327
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007327 25 mg 25mg
30009813 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007327 Supplier Toronto Research Chemicals Supplier No. TRCB69147525MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 159351-68-5
Molecular Formula C59 H97 N O14 Si
Molecular Weight (g/mol) 1072.48
Synonym 42-O-[2-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]ethyl]rapamycin, 23,27-Epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclohentriacontine Rapamycin Deriv., Everolimus tert-Butyldimethylsilyl Ether
IUPAC Name (1R,9S,12S,15R,16E,18R,19R,21R,23S,24E,26E,28E,30S,32S,35R)-12-[(2R)-1-[(1S,3R,4R)-4-[2-[tert-butyl(dimethyl)silyl]oxyethoxy]-3-methoxycyclohexyl]propan-2-yl]-1,18-dihydroxy-19,30-dimethoxy-15,17,21,23,29,35-hexamethyl-11,36-dioxa-4-azatricyclo[30.3.1.04,9]hexatriaconta-16,24,26,28-tetraene-2,3,10,14,20-pentone
SMILES CO[C@@H]1C[C@H](C[C@@H](C)[C@@H]2CC(=O)[C@H](C)\C=C(/C)\[C@@H](O)[C@@H](OC)C(=O)[C@H](C)C[C@H](C)\C=C\C=C\C=C(/C)\[C@H](C[C@@H]3CC[C@@H](C)[C@@](O)(O3)C(=O)C(=O)N4CCCC[C@H]4C(=O)O2)OC)CC[C@H]1OCCO[Si](C)(C)C(C)(C)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C59H97NO14Si/c1-37-21-17-16-18-22-38(2)49(68-11)35-45-26-24-43(7)59(67,74-45)55(64)56(65)60-28-20-19-23-46(60)57(66)73-50(36-47(61)39(3)32-42(6)53(63)54(70-13)52(62)41(5)31-37)40(4)33-44-25-27-48(51(34-44)69-12)71-29-30-72-75(14,15)58(8,9)10/h16-18,21-22,32,37,39-41,43-46,48-51,53-54,63,67H,19-20,23-31,33-36H2,1-15H3/b18-16+,21-17+,38-22+,42-32+/t37-,39-,40-,41-,43-,44+,45+,46+,48-,49+,50+,51-,53-,54+,59-/m1/s1
Formula Weight 1071.6678
Chemical Name or Material 42-O-tert-Butyldimethylsilyloxyethyl Rapamycin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.