Learn More
Skytec™ 4530 Disposable 3-Panel Fold Flat Respirator - FFP3
Product Code. 30242987 Shop All Skytec Products
Change view
Click to view available options
Quantity:
20/Pk.
Unit Size:
Pack of 20
Product Code. Quantity Unit Size
30242987 20/Pk. Pack of 20
1 options
Product Code. 30242987 Supplier Skytec™ Supplier No. ASRAPL300AZ
  • ONLINE EXCLUSIVE PRICE
    162.56 €
    108.27 € / Pack of 20
    Save 54.29 €
    33% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
The 4500 Series is the newest addition to Skytec's range of premium respirators. Featuring premium European-Made filter media across valved and unvalved variants, the 4500 Series range provides low breathing resistance and excellent Face Fit Factor.
  • Premium disposable fold flat respirator for easy storage, donning and doffing
  • Secure, metal-free harness system with color coded textile elastic for easy identification of FFP level
  • Soft, flexible and adjustable nose bridge for increased fit factor and comfort
  • Multilayer electrostatic filter media for high-performing filtration with low breathing resistance
  • Full-width internal faceseal, made from soft touch PVC for high comfort
  • Ergonomic design for a secure fit on a wide range of face shapes
  • High flow exhalation valve for low breathing resistance, color-coded for easy identification of FFP level
  • Contoured noseseal for high compatibility with other types of PPE
  • All variants complaint to EN 149:2001+A1:2009. Unvalved variant compliant to EN 14683:2019 for use in healthcare
  • Latex-free and low-linting for use in a wide range of environments
  • UK designed, developed and manufactured.
Specifications
Assigned Protection Factor (APF) 20
Certifications/Compliance CE III , UKCA , UK Medical Device Directive: 93/42/EEC and 2007/47/EC , EU Medical Device Regulation: 2017/745 , EN 149:2000 + A1:2009 FFP3 NR , EN 14683:2019 Type IIR
Color White
Design Type Flat-fold
Disposable Single-use
Filter Type FFP3
Material Polypropylene
Product Type Healthcare Particulate Respirator and Surgical Mask
Size One Size
With Exhalation Valve No
Filtration Layer Material Non-woven Textile
Material Headstrap Latex-Free Elastane,Polyester,Textile Elastic
Model 4500
Nose Piece Material Polyethylene Foam
Quantity 20/Pk.
Sterility Non-sterile