missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates™ pureGrade™ Strip Plates

Product Code. 11633189 Shop All Brand Products
Change view
Click to view available options
Total Volume (Metric) Well:
350 μL
360 μL
Breakapart:
No
Yes
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Total Volume (Metric) Well Breakapart unitSize
11633189 360 μL No Pack of 100
11643189 350 μL Yes Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11633189 Supplier BRAND™ Supplier No. 782300
Discount Lab Supplies
Use promo code 28170
View all promotions
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Non-treated surface strip plates designed to maximize sample recovery and minimize cross-contamination.

  • Medium binding and non-sterile
  • Transparent with flat bottoms
  • Available with or without grid
  • Free from endotoxins, DNase, DNA, RNase, and non-cytotoxic

With grid: Individual strips can be removed from the plate and reinserted. Strips can also be divided into 8 individual vessels and inserted separately into the frame plate.

Without grid: Individual strips can be removed from the frame and reinserted. Strips are not divisible.

Applications: Ideal for homogeneous assays, screening and storage.

TRUSTED_SUSTAINABILITY

Specifications

Barcode Alphanumeric
Breakapart No
Binding Property Untreated
Certified Life Science Quality
Format Strip Plate
Array 12 x 8
Autoclavable Not Autoclavable
Binding Type Medium Binding
Color Clear
Disposable Single-use
Product Type Strip plates
For Use With (Equipment) Microplate readers
Material Polystyrene
Volume (Metric) Well 360 μL
No. of Wells 96
Volume (Metric) Working 360 μL
Sterility Non-sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 360 μL
Well Shape Flat Bottom (F-well)
Dnase Free Yes
For Use With (Application) Homogeneous assays, screenings, and storage
Includes 96-well strip plate
Quantity 100/Pk.
RNase Free Yes
Target Molecule Immunoglobulin G (IgG)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.